Learn More
5-Fluoro-2-methoxyaniline, 97%, Thermo Scientific™
Catalog Number 15435257
Click to view available options
Quantity:
250 mg
1 g
1978-39-8
C7H8FNO
141.15
MFCD00077537
VYZUBHRSGQAROM-UHFFFAOYSA-N
2-amino-4-fluoroanisole, 5-fluoro-2-methoxyphenylamine, 5-fluoro-o-anisidine, 5-fluoro-2-methoxybenzenamine, 3-fluoro-6-methoxyaniline, benzenamine, 5-fluoro-2-methoxy, 2-amino-4-fluoro-1-methoxybenzene, 4-fluoro-2-aminoanisole, 2-methoxy-5-fluoroaniline, 2-methoxyl-5-fluoroaniline
223105
5-fluoro-2-methoxyaniline
COC1=CC=C(F)C=C1N
1978-39-8
C7H8FNO
141.15
MFCD00077537
VYZUBHRSGQAROM-UHFFFAOYSA-N
2-amino-4-fluoroanisole, 5-fluoro-2-methoxyphenylamine, 5-fluoro-o-anisidine, 5-fluoro-2-methoxybenzenamine, 3-fluoro-6-methoxyaniline, benzenamine, 5-fluoro-2-methoxy, 2-amino-4-fluoro-1-methoxybenzene, 4-fluoro-2-aminoanisole, 2-methoxy-5-fluoroaniline, 2-methoxyl-5-fluoroaniline
223105
5-fluoro-2-methoxyaniline
COC1=CC=C(F)C=C1N
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Refractive Index 1.543
Quantity 250 mg
UN Number UN2810
Formula Weight 141.15
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-methoxyaniline