Få mere at vide
Molecular Probes™ 5-FAM (5-Carboxyfluorescein), single isomer
Mængde:
100 mg
Pakningsstørrelse:
100mg
Beskrivelse
The single isomer, 5-FAM, contains a carboxylic acid that can be used to react with primary amines via carbodiimide activation of the carboxylic acid. Fluorescein is the most common fluorescent derivatization reagent for labeling biomolecules. In addition to its relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility, fluorescein has an excitation maximum that closely matches the 488 nm spectral line of the argon-ion laser.
Tekniske data
Tekniske data
| Mængde | 100 mg |
| Produkttype | 5-FAM |
| Indhold og opbevaring | Store at room temperature and protect from light. |
| Kemisk reaktivitet | Amine |
| Forsendelsestilstand | Room Temperature |
| Reaktiv del | Carboxylic Acid |
| Etiket eller farvestof | FITC (Fluorescein) |
| Etikettype | Classic Dyes |
| Excitation | 488 |