Få mere at vide
5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanone-d3, TRC
Artikelnummer. 30373245
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30373245 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30373245 Leverandør Toronto Research Chemicals Leverandørnr. TRCE91900750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanone-d3
CAS 156420-67-6
Molekylær formel C7 D3 H7 O3
Mængde 50 mg
Synonym 5-(ethyl-2,2,2-d3)-3-hydroxy-4-methyl-2(5H)-furanone
InChI formel InChI=1S/C7H10O3/c1-3-5-4(2)6(8)7(9)10-5/h5,8H,3H2,1-2H3/i1D3
SMIL [2H]C([2H])([2H])CC1OC(=O)C(=C1C)O
IUPAC navn 4-hydroxy-3-methyl-2-(2,2,2-trideuterioethyl)-2H-furan-5-one
Molekylvægt (g/mol) 145.171
Formel vægt 145.082