Få mere at vide
5-Ethyl-2-pyridineethanol Tosylate, TRC
Artikelnummer. 30379847
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30379847 500 mg
30379368 100 mg
30379364 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30379847 Leverandør Toronto Research Chemicals Leverandørnr. TRCE925555500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 144809-27-8
Molekylær formel C16H19NO3S
Molekylvægt (g/mol) 305.39
Synonym 5-Ethyl-2-pyridineethanol 2-(4-Methylbenzenesulfonate), 2-(5-Ethyl-2-pyridyl)ethyl Tosylate, Toluene-4-sulfonic Acid 2-(5-Ethylpyridin-2-yl)ethyl Ester
IUPAC navn 2-(5-ethylpyridin-2-yl)ethyl 4-methylbenzenesulfonate
SMIL CCc1ccc(CCOS(=O)(=O)c2ccc(C)cc2)nc1
Tekniske data
Kemisk navn eller materiale 5-Ethyl-2-pyridineethanol Tosylate
Mængde 500 mg
InChI formel InChI=1S/C16H19NO3S/c1-3-14-6-7-15(17-12-14)10-11-20-21(18,19)16-8-4-13(2)5-9-16/h4-9,12H,3,10-11H2,1-2H3
Formel vægt 305.1086