Få mere at vide
5-Ethyl-2-[2-(4-nitrophenoxy)ethyl]pyridine, TRC
Artikelnummer. 30379290
Skift visning
Mængde:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30379290 25 g
30371977 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30379290 Leverandør Toronto Research Chemicals Leverandørnr. TRCE92522525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 85583-54-6
Molekylær formel C15H16N2O3
Molekylvægt (g/mol) 272.3
Synonym 4-[2-(5-Ethyl-2-pyridinyl)ethoxy]nitrobenzene
SMIL CCc1ccc(CCOc2ccc(cc2)[N+](=O)[O-])nc1
Tekniske data
Kemisk navn eller materiale 5-Ethyl-2-[2-(4-nitrophenoxy)ethyl]pyridine
Mængde 25 g
InChI formel InChI=1S/C15H16N2O3/c1-2-12-3-4-13(16-11-12)9-10-20-15-7-5-14(6-8-15)17(18)19/h3-8,11H,2,9-10H2,1H3
Formel vægt 272.1161