Få mere at vide
5-Ethyl-1,3-benzenediol, TRC
Artikelnummer. 30137079
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137079 1 g
30138454 500 mg
30010478 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137079 Leverandør Toronto Research Chemicals Leverandørnr. TRCE8967001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4299-72-3
Molekylær formel C8H10O2
Molekylvægt (g/mol) 138.16
Synonym 5-Ethyl-resorcinol, 5-Ethylresorcin, 5-Ethylresorcinol, Homoorcinol;
SMIL CCc1cc(O)cc(O)c1
Tekniske data
Kemisk navn eller materiale 5-Ethyl-1,3-benzenediol
Mængde 1 g
InChI formel InChI=1S/C8H10O2/c1-2-6-3-7(9)5-8(10)4-6/h3-5,9-10H,2H2,1H3
Formel vægt 138.0681