Få mere at vide
5-Deschlorolifitegrast, TRC
Artikelnummer. 30180050
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180050 1 mg
30179943 10 mg
30179875 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30180050 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2216501MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2322380-15-2
Molekylær formel C29H25ClN2O7S
Molekylvægt (g/mol) 581.04
Synonym (S)-2-(2-(Benzofuran-6-carbonyl)-5-chloro-1,2,3,4-tetrahydroisoquinoline-6-carboxamido)-3-(3-(methylsulfonyl)phenyl)propanoic Acid
IUPAC navn (2S)-2-[[2-(1-benzofuran-6-carbonyl)-5-chloro-3,4-dihydro-1H-isoquinoline-6-carbonyl]amino]-3-(3-methylsulfonylphenyl)propanoic acid
SMIL CS(=O)(=O)c1cccc(C[C@H](NC(=O)c2ccc3CN(CCc3c2Cl)C(=O)c4ccc5ccoc5c4)C(=O)O)c1
Tekniske data
Kemisk navn eller materiale 5-Deschlorolifitegrast
Mængde 1 mg
InChI formel InChI=1S/C29H25ClN2O7S/c1-40(37,38)21-4-2-3-17(13-21)14-24(29(35)36)31-27(33)23-8-7-20-16-32(11-9-22(20)26(23)30)28(34)19-6-5-18-10-12-39-25(18)15-19/h2-8,10,12-13,15,24H,9,11,14,16H2,1H3,(H,31,33)(H,35,36)/t24-/m0/s1
Formel vægt 580.1071