missing translation for 'onlineSavingsMsg'
Få mere at vide

5’-Deoxy-5-fluoro Cytidine, TRC

Artikelnummer. 30132677
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132677 500 mg
30134172 100 mg
30004616 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30132677 Leverandør Toronto Research Chemicals Leverandørnr. TRCD232950500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66335-38-4
Molekylær formel C9 H12 F N3 O4
Molekylvægt (g/mol) 245.21
Synonym 5'-deoxy-5-fluorocytidine, 5'-DFCR, 5'-Deoxy-5-fluorocytidine, Capecitabine Imp. A (EP), Capecitabine Related Compound A (USP)
IUPAC navn 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoropyrimidin-2-one
SMIL C[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=C(F)C(=NC2=O)N

Specifications

Kemisk navn eller materiale 5'-Deoxy-5-fluorocytidine
Mængde 500 mg
InChI formel InChI=1S/C9H12FN3O4/c1-3-5(14)6(15)8(17-3)13-2-4(10)7(11)12-9(13)16/h2-3,5-6,8,14-15H,1H3,(H2,11,12,16)/t3-,5-,6-,8-/m1/s1
Formel vægt 245.0812
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.