Få mere at vide
5-Dehydro Tolvaptan-d7, TRC
Artikelnummer. 30352824
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30352824 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30352824 Leverandør Toronto Research Chemicals Leverandørnr. TRCD23035210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Dehydro Tolvaptan-d7
CAS 1296212-29-7
Molekylær formel C26H16D7ClN2O3
Mængde 10 mg
Synonym N-[4-[(7-Chloro-2,3,4,5-tetrahydro-5-oxo-1H-1-benzazepin-1-yl)carbonyl]-3-methylphenyl]-2-methylbenzamide-d7, 1H-1-Benzazepine, Benzamide Deriv.-d7, 7-Chloro-1-[2-methyl-4-[(2-methylbenzoyl)amino]benzoyl]-5-Oxo-2,3,4,5-tetrahydro-1H-1-benzazepine-d7
InChI formel InChI=1S/C26H23ClN2O3/c1-16-6-3-4-7-20(16)25(31)28-19-10-11-21(17(2)14-19)26(32)29-13-5-8-24(30)22-15-18(27)9-12-23(22)29/h3-4,6-7,9-12,14-15H,5,8,13H2,1-2H3,(H,28,31)/i1D3,3D,4D,6D,7D
SMIL [2H]c1c([2H])c([2H])c(c(C(=O)Nc2ccc(C(=O)N3CCCC(=O)c4cc(Cl)ccc34)c(C)c2)c1[2H])C([2H])([2H])[2H]
IUPAC navn N-[4-(7-chloro-5-oxo-3,4-dihydro-2H-1-benzazepine-1-carbonyl)-3-methylphenyl]-2,3,4,5-tetradeuterio-6-(trideuteriomethyl)benzamide
Molekylvægt (g/mol) 453.97
Formel vægt 453.1837