Learn More
5-Cyclopropyl-1,2,4-oxadiazol-3-amine, 97%, Thermo Scientific™
Catalog Number p-7077336
Click to view available options
Quantity:
1 g
5 g
868696-42-8
C5H7N3O
125.13
OHSZRALIDGBNES-UHFFFAOYSA-N
1,2,4-oxadiazol-3-amine, 5-cyclopropyl
51072255
5-cyclopropyl-1,2,4-oxadiazol-3-amine
C1CC1C2=NC(=NO2)N
868696-42-8
C5H7N3O
125.13
OHSZRALIDGBNES-UHFFFAOYSA-N
1,2,4-oxadiazol-3-amine, 5-cyclopropyl
51072255
5-cyclopropyl-1,2,4-oxadiazol-3-amine
C1CC1C2=NC(=NO2)N
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 125.13
Percent Purity 97%
Chemical Name or Material 5-Cyclopropyl-1,2,4-oxadiazol-3-amine
compliance-icons
Product Identifier
  • 5-Cyclopropyl-1,2,4-oxadiazol-3-amine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.