Få mere at vide
5-Cyanopentanamide, TRC
Artikelnummer. 30363019
Skift visning
Mængde:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30363019 5 g
30368185 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30363019 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9876205G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2304-58-7
Molekylær formel C6 H10 N2 O
Molekylvægt (g/mol) 126.16
Synonym 5-Cyanovaleramide
IUPAC navn 5-cyanopentanamide
SMIL NC(=O)CCCCC#N
Tekniske data
Kemisk navn eller materiale 5-Cyanopentanamide
Mængde 5 g
InChI formel InChI=1S/C6H10N2O/c7-5-3-1-2-4-6(8)9/h1-4H2,(H2,8,9)
Formel vægt 126.0793