Learn More
Alfa Aesar™ 5-Cyano-3-(3-oxocyclopentyl)indole, 95%
Brand: Alfa Aesar™ H63625.03
Additional Details : CAS Number : 675831-64-8 Weight : 0.00100kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III
Quantity | 1g |
---|
Chemical Identifiers
675831-64-8 | |
224.263 | |
3-3-oxocyclopentyl-1h-indole-5-carbonitrile, 3-3-oxocyclopentyl-5-cyano-1h-indole, 3-3-oxo-cyclopentyl-1h-indole-5-carbonitrile, 1h-indole-5-carbonitrile, 3-3-oxocyclopentyl | |
3-(3-oxocyclopentyl)-1H-indole-5-carbonitrile |
C14H12N2O | |
PCCIEBVCPXEIFS-UHFFFAOYSA-N | |
22139459 | |
C1CC(=O)CC1C2=CNC3=C2C=C(C=C3)C#N |
Specifications
5-Cyano-3-(3-oxocyclopentyl)indole | |
675831-64-8 | |
UN3439 | |
PCCIEBVCPXEIFS-UHFFFAOYSA-N | |
3-(3-oxocyclopentyl)-1H-indole-5-carbonitrile | |
22139459 | |
Solid |
1g | |
C14H12N2O | |
3-3-oxocyclopentyl-1h-indole-5-carbonitrile, 3-3-oxocyclopentyl-5-cyano-1h-indole, 3-3-oxo-cyclopentyl-1h-indole-5-carbonitrile, 1h-indole-5-carbonitrile, 3-3-oxocyclopentyl | |
C1CC(=O)CC1C2=CNC3=C2C=C(C=C3)C#N | |
224.263 | |
224.26 | |
95% |