Få mere at vide
5-Cholesten-24(RS)-ethyl-3beta-ol-25,26,26,26,27,27,27-d7, CDN
Artikelnummer. 30310148
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30310148 0.01 g
30319237 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310148 Leverandør CDN Leverandørnr. CDND63870.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale beta-Sitosterol-d7 (mixture of diasteromers)
CAS 2260669-28-9
Molekylær formel C29 D7 H43 O
Mængde 0.01 g
Synonym 5-Cholesten-24(RS)-ethyl-3β-ol-25,26,26,26,27,27,27-d7, 24(R,S)-Stigmast-5-en-3β-ol;
InChI formel InChI=1 S/C29H50O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h10,19-21,23-27,30 H,7-9,11-18H2,1-6H3/t20-,21?,23+,24+,25-,26+,27+,28+,29-/m1/s1/i2D3,3D3,19 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(C(CC)CC[C@@H](C)[C@H]1 CC[C@H]2 [C@@H]3 CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3 CC[C@]12 C)C([2 H])([2 H])[2 H]
IUPAC navn (3 S,8 S,9 S,10 R,13 R,14 S,17 R)-10,13-dimethyl-17-[(2 R)-6,7,7,7-tetradeuterio-5-ethyl-6-(trideuteriomethyl)heptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1 H-cyclopenta[a]phenanthren-3-ol
Molekylvægt (g/mol) 421.75
Formel vægt 421.4301 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity