Få mere at vide
5-Cholenic Acid-3Beta-ol, TRC
Artikelnummer. 30138169
Skift visning
Mængde:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138169 10 mg
30137064 250 mg
30006928 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138169 Leverandør Toronto Research Chemicals Leverandørnr. TRCC43101010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5255-17-4
Molekylær formel C24 H38 O3
Molekylvægt (g/mol) 374.56
Synonym 3β-Hydroxychol-5-en-24-oic Acid, 3β-Hydroxychol-5-enic Acid, 3β-Hydroxy-5-cholenic Acid, 3β-Hydroxy-δ5-cholenic Acid, 3β-Hydroxychol-5-en-24-ic Acid, 3β-Hydroxychol-5-enoic Acid, 3β-Hydroxycholenoic Acid, Cholenic Acid, ZINC 04831336, δ5-3β-Hydroxycholenic Acid, δ5-Cholenic Acid
IUPAC navn (4R)-4-[(3S,8S,9S,10R,13R,14S,17R)-3-hydroxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMIL C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale 5-Cholenic Acid-3Beta-ol
Mængde 10 mg
InChI formel InChI=1S/C24H38O3/c1-15(4-9-22(26)27)19-7-8-20-18-6-5-16-14-17(25)10-12-23(16,2)21(18)11-13-24(19,20)3/h5,15,17-21,25H,4,6-14H2,1-3H3,(H,26,27)/t15-,17+,18+,19-,20+,21+,23+,24-/m1/s1
Formel vægt 374.2821