Få mere at vide
5-(Chlorosulfonyl)-3-methylthiophene-2-carboxylic Acid, TRC
Artikelnummer. 30367011
Skift visning
Mængde:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367011 250 mg
30360475 25 mg
30354749 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30367011 Leverandør Toronto Research Chemicals Leverandørnr. TRCC375673250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1114823-93-6
Molekylær formel C6H5O4S2Cl
Molekylvægt (g/mol) 240.68
IUPAC navn 5-chlorosulfonyl-3-methylthiophene-2-carboxylic acid
SMIL Cc1cc(sc1C(=O)O)S(=O)(=O)Cl
Tekniske data
Kemisk navn eller materiale 5-(Chlorosulfonyl)-3-methylthiophene-2-carboxylic Acid
Mængde 250 mg
InChI formel InChI=1S/C6H5ClO4S2/c1-3-2-4(13(7,10)11)12-5(3)6(8)9/h2H,1H3,(H,8,9)
Formel vægt 239.9318