Få mere at vide
5-Chlorosulfonyl-2-methoxybenzoic Acid-d3, TRC
Artikelnummer. 30365432
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30365432 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30365432 Leverandør Toronto Research Chemicals Leverandørnr. TRCC31521725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Chlorosulfonyl-2-methoxybenzoic Acid-d3
CAS 123958-84-9
Molekylær formel C8H4D3ClO5S
Mængde 25 mg
Synonym 2-Methoxy-5-chlorosulfonylbenzoic Acid-d3, 3-Carboxy-4-methoxybenzenesulfonyl-d3 Chloride, 5-(Chlorosulfonyl)-2-(methyloxy)benzoic Acid-d3, 5-(Chlorosulfonyl)-o-anisic Acid-d3
InChI formel InChI=1S/C8H7ClO5S/c1-14-7-3-2-5(15(9,12)13)4-6(7)8(10)11/h2-4H,1H3,(H,10,11)/i1D3
SMIL [2H]C([2H])([2H])Oc1ccc(cc1C(=O)O)S(=O)(=O)Cl
IUPAC navn 5-chlorosulfonyl-2-(trideuteriomethoxy)benzoic acid
Molekylvægt (g/mol) 253.67
Formel vægt 252.9891