Få mere at vide
5-(Chloropyridin-2-yl-carbamoyl)pyrazine, Mikromol™
Artikelnummer. 17943216
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17943216 100 mg
17973876 25 mg
30459558 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17943216 Leverandør Mikromol Leverandørnr. MM0149.07
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-(Chloropyridin-2-yl-carbamoyl)pyrazine
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-(Chloropyridin-2-yl-carbamoyl)pyrazine
CAS 349125-10-6
Analyt- eller komponentnavne 5-(Chloropyridin-2-yl-carbamoyl)pyrazine
Forsendelsestilstand Room Temperature
Molekylær formel C10 H7 Cl N4 O
Certificeringer/compliance ISO 9001
Synonym 5-(Chloropyridin-2-yl-carbamoyl)pyrazine
InChI formel InChI=1S/C10H7ClN4O/c11-7-1-2-9(14-5-7)15-10(16)8-6-12-3-4-13-8/h1-6H,(H,14,15,16)
SMIL Clc1ccc(NC(=O)c2cnccn2)nc1
IUPAC navn N-(5-chloropyridin-2-yl)pyrazine-2-carboxamide
Molekylvægt (g/mol) 234.64
Mængde 100 mg
Formel vægt 234.0308 g/mol