Få mere at vide
5-(Chloromethyl)-2-hydroxybenzaldehyde, TRC
Artikelnummer. 30363005
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30363005 100 mg
30351252 50 mg
30358091 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30363005 Leverandør Toronto Research Chemicals Leverandørnr. TRCC413400100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 23731-06-8
Molekylær formel C8H7O2Cl
Molekylvægt (g/mol) 170.59
IUPAC navn 5-(chloromethyl)-2-hydroxybenzaldehyde
SMIL Oc1ccc(CCl)cc1C=O
Tekniske data
Kemisk navn eller materiale 5-(Chloromethyl)-2-hydroxybenzaldehyde
Mængde 100 mg
InChI formel InChI=1S/C8H7ClO2/c9-4-6-1-2-8(11)7(3-6)5-10/h1-3,5,11H,4H2
Formel vægt 170.0135