missing translation for 'onlineSavingsMsg'
Få mere at vide

5-(Chloromethyl)-2-hydroxybenzaldehyde, TRC

Artikelnummer. 30351252
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30351252 50 mg
30358091 500 mg
30363005 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30351252 Leverandør Toronto Research Chemicals Leverandørnr. TRCC41340050MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 23731-06-8
Molekylær formel C8H7O2Cl
Molekylvægt (g/mol) 170.59
IUPAC navn 5-(chloromethyl)-2-hydroxybenzaldehyde
SMIL Oc1ccc(CCl)cc1C=O

Specifications

Kemisk navn eller materiale 5-(Chloromethyl)-2-hydroxybenzaldehyde
Mængde 50 mg
InChI formel InChI=1S/C8H7ClO2/c9-4-6-1-2-8(11)7(3-6)5-10/h1-3,5,11H,4H2
Formel vægt 170.0135
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.