Få mere at vide
5-Chlorodescyano Citalopram Hydrobromide, TRC
Artikelnummer. 30365275
Skift visning
Mængde:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30365275 250 mg
30350067 100 mg
30365583 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30365275 Leverandør Toronto Research Chemicals Leverandørnr. TRCC366035250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1332724-08-9
Molekylær formel C19 H21 Cl F N O . Br H
Molekylvægt (g/mol) 414.74
Synonym 1-Isobenzofuranpropanamine, 5-chloro-1-(4-fluorophenyl)-1,3-dihydro-N,N-dimethyl-, hydrobromide (1:1), 1-(4'-fluorophenyl)-1-(3-dimethylaminopropyl)-5-chlorophthalane hydrobromide, 3-[(1RS)-5-Chloro-1(4-fluorophenyl)-1,3-dihydroisobenzofuran-1-yl]-N,N-dimethylpropan-1-amine Hydrobromide
IUPAC navn 3-[5-chloro-1-(4-fluorophenyl)-3H-2-benzofuran-1-yl]-N,N-dimethylpropan-1-amine;hydrobromide
SMIL Br.CN(C)CCCC1(OCc2cc(Cl)ccc12)c3ccc(F)cc3
Tekniske data
Kemisk navn eller materiale 3-[(1RS)-5-Chloro-1(4-fluorophenyl)-1,3-dihydroisobenzofuran-1-yl]-N,N-dimethylpropan-1-amine Hydrobromide
Mængde 250 mg
InChI formel InChI=1S/C19H21ClFNO.BrH/c1-22(2)11-3-10-19(15-4-7-17(21)8-5-15)18-9-6-16(20)12-14(18)13-23-19;/h4-9,12H,3,10-11,13H2,1-2H3;1H
Formel vægt 413.0557