Få mere at vide
5-Chloroacetyl-6-chloro-1,3-dihydroindol-2H-indone-2-one, TRC
Artikelnummer. 30358976
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30358976 1 g
30352329 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30358976 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3633701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 118307-04-3
Molekylær formel C10 H7 Cl2 N O2
Molekylvægt (g/mol) 244.07
Synonym 6-Chloro-5-(2-chloroacetyl)-1,3-dihydro-2H-indol-2-one, 2H-Indol-2-one, 6-chloro-5-(chloroacetyl)-1,3-dihydro- (9CI), 5-(2-Chloroacetyl)-6-chloro-2-oxindole, 5-(2-Chloroacetyl)-6-chlorooxindole, 6-Chloro-5-(2-chloroacetyl)-1,3-dihydro-2H-indol-2-one, 6-Chloro-5-(2-chloroacetyl)indolin-2-one, 6-Chloro-5-(chloroacetyl)oxindole, 6-Chloro-5-chloroacetyl-2-oxoindoline
IUPAC navn 6-chloro-5-(2-chloroacetyl)-1,3-dihydroindol-2-one
SMIL ClCC(=O)c1cc2CC(=O)Nc2cc1Cl
Tekniske data
Kemisk navn eller materiale 5-Chloroacetyl-6-chloro-1,3-dihydroindol-2H-indone-2-one
Mængde 1 g
InChI formel InChI=1S/C10H7Cl2NO2/c11-4-9(14)6-1-5-2-10(15)13-8(5)3-7(6)12/h1,3H,2,4H2,(H,13,15)
Formel vægt 242.9854