Få mere at vide
5-Chloroacenaphthenone, TRC
Artikelnummer. 30354354
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30354354 50 mg
30365482 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30354354 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36706550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 37568-51-7
Molekylær formel C12H7ClO
Molekylvægt (g/mol) 202.64
Synonym 5-Chloro-1(2H)-acenaphthylenone
IUPAC navn 5-chloro-2H-acenaphthylen-1-one
SMIL Clc1ccc2CC(=O)c3cccc1c23
Tekniske data
Kemisk navn eller materiale 5-Chloroacenaphthenone
Mængde 50 mg
InChI formel InChI=1S/C12H7ClO/c13-10-5-4-7-6-11(14)9-3-1-2-8(10)12(7)9/h1-5H,6H2
Formel vægt 202.0185