Få mere at vide
5-Chloro-N1-phenylbenzene-1,2-diamine, TRC
Artikelnummer. 30362726
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362726 100 mg
30353504 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30362726 Leverandør Toronto Research Chemicals Leverandørnr. TRCC374850100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Chloro-N1-phenylbenzene-1,2-diamine
CAS 68406-47-3
Molekylær formel C12H11ClN2
Mængde 100 mg
Synonym 2-Amino-5-Chloro-N-phenylaniline, 2-Amino-5-chlorodiphenylamine, 4-Chloro-N'-phenylbenzene-1,2-diamine, 4-Chloro-N2-phenyl-o-phenylenediamine
InChI formel InChI=1S/C12H11ClN2/c13-9-6-7-11(14)12(8-9)15-10-4-2-1-3-5-10/h1-8,15H,14H2
SMIL NC1=CC=C(Cl)C=C1NC2=CC=CC=C2
IUPAC navn 5-chloro-N1-phenylbenzene-1,2-diamine
Molekylvægt (g/mol) 218.68
Formel vægt 218.06