Få mere at vide
5-Chloro-N-methylthiophene-2-carboxamide, TRC
Artikelnummer. 30368507
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30368507 1 g
30364015 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30368507 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3738401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 97799-98-9
Molekylær formel C6 H6 Cl N O S
Molekylvægt (g/mol) 175.64
Synonym 2-Thiophenecarboxamide, 5-chloro-N-methyl- (6CI, 9CI, ACI), 5-Chloro-N-methyl-2-thiophenecarboxamide (ACI), 5-Chloro-N-methyl-2-thiophenecarboxamide, 5-Chloro-N-methyl-2-thiophenecarboxamide
IUPAC navn 5-chloro-N-methylthiophene-2-carboxamide
SMIL CNC(=O)c1ccc(Cl)s1
Tekniske data
Kemisk navn eller materiale 5-Chloro-N-methylthiophene-2-carboxamide
Mængde 1 g
InChI formel InChI=1S/C6H6ClNOS/c1-8-6(9)4-2-3-5(7)10-4/h2-3H,1H3,(H,8,9)
Formel vægt 174.9859