Få mere at vide
5-Chloro Imidacloprid, TRC
Artikelnummer. 30136859
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136859 100 mg
30003684 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136859 Leverandør Toronto Research Chemicals Leverandørnr. TRCC366910100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 135769-74-3
Molekylær formel C9 H9 Cl2 N5 O2
Molekylvægt (g/mol) 290.11
Synonym 1H-Imidazol-2-amine, 1-[(5,6-dichloro-3-pyridinyl)methyl]-4,5-dihydro-N-nitro-, 1-[(5,6-Dichloro-3-pyridinyl)methyl]-4,5-dihydro-N-nitro-1H-imidazol-2-amine, N-[1-[(5,6-Dichloro-3-pyridyl)methyl]-4,5-dihydroimidazol-2-yl]nitramide, 5-Chloro Imidacloprid, 5-Chloroimidacloprid
IUPAC navn N-[1-[(5,6-dichloropyridin-3-yl)methyl]-4,5-dihydroimidazol-2-yl]nitramide
SMIL [O-][N+](=O)NC1=NCCN1Cc2cnc(Cl)c(Cl)c2
Tekniske data
Kemisk navn eller materiale 5-Chloro Imidacloprid
Mængde 100 mg
InChI formel InChI=1S/C9H9Cl2N5O2/c10-7-3-6(4-13-8(7)11)5-15-2-1-12-9(15)14-16(17)18/h3-4H,1-2,5H2,(H,12,14)
Formel vægt 289.0133