Få mere at vide
5-Chloro-DL-tryptophan, TRC
Artikelnummer. 30368446
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30368446 50 mg
30366233 100 mg
30357366 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30368446 Leverandør Toronto Research Chemicals Leverandørnr. TRCC42322050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 154-07-4
Molekylær formel C11H11ClN2O2
Molekylvægt (g/mol) 238.67
IUPAC navn 2-amino-3-(5-chloro-1H-indol-3-yl)propanoic acid
SMIL NC(Cc1c[nH]c2ccc(Cl)cc12)C(=O)O
Tekniske data
Kemisk navn eller materiale 5-Chloro-DL-tryptophan
Mængde 50 mg
InChI formel InChI=1S/C11H11ClN2O2/c12-7-1-2-10-8(4-7)6(5-14-10)3-9(13)11(15)16/h1-2,4-5,9,14H,3,13H2,(H,15,16)
Formel vægt 238.0509