Få mere at vide
5-Chloro-8-methylquinoline, TRC
Artikelnummer. 30356257
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30356257 100 mg
30361075 250 mg
30361246 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30356257 Leverandør Toronto Research Chemicals Leverandørnr. TRCC384828100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 78941-95-4
Molekylær formel C10H8NCl
Molekylvægt (g/mol) 177.63
IUPAC navn 5-chloro-8-methylquinoline
SMIL Cc1ccc(Cl)c2cccnc12
Tekniske data
Kemisk navn eller materiale 5-Chloro-8-methylquinoline
Mængde 100 mg
InChI formel InChI=1S/C10H8ClN/c1-7-4-5-9(11)8-3-2-6-12-10(7)8/h2-6H,1H3
Formel vægt 177.0345