missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-4-azaindole-2-carboxylic acid, 98%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Used for pharmaceutical and cytokine signaling intermediates.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
Used for pharmaceutical and cytokine signaling intermediates.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
Spezifikation
Melting Point | ∽295°C (decomposition) |
Quantity | 250 mg |
Solubility Information | Insoluble in water. |
Formula Weight | 196.59 |
Percent Purity | 98% |
Chemical Name or Material | 5-Chloro-4-azaindole-2-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'