missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-4-azaindole-2-carboxylic acid, 98%, Thermo Scientific™

Catalog Number 15426477
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
800401-63-2
C8H5ClN2O2
196.59
MFCD18261123
LTAXMSVURJPVQV-UHFFFAOYSA-N
5-chloro-1h-pyrrolo 3,2-b pyridine-2-carboxylic acid, 1h-pyrrolo 3,2-b pyridine-2-carboxylic acid, 5-chloro, 5-chloro-4-azaindole-2-carboxylic acid, chloropyrrolobpyridinecarboxylicacid, 5-chloro-1h-pyrrolo 2,3-pyridine-2-carboxylic acid
11665585
5-chloro-1H-pyrrolo[3,2-b]pyridine-2-carboxylic acid
C1=CC(=NC2=C1NC(=C2)C(=O)O)Cl
This item is not returnable. View return policy
800401-63-2
C8H5ClN2O2
196.59
MFCD18261123
LTAXMSVURJPVQV-UHFFFAOYSA-N
5-chloro-1h-pyrrolo 3,2-b pyridine-2-carboxylic acid, 1h-pyrrolo 3,2-b pyridine-2-carboxylic acid, 5-chloro, 5-chloro-4-azaindole-2-carboxylic acid, chloropyrrolobpyridinecarboxylicacid, 5-chloro-1h-pyrrolo 2,3-pyridine-2-carboxylic acid
11665585
5-chloro-1H-pyrrolo[3,2-b]pyridine-2-carboxylic acid
C1=CC(=NC2=C1NC(=C2)C(=O)O)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used for pharmaceutical and cytokine signaling intermediates.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point ∽295°C (decomposition)
Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 196.59
Percent Purity 98%
Chemical Name or Material 5-Chloro-4-azaindole-2-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'