missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-3-phenyl-2,1-benzisoxazole, 98+%, Thermo Scientific™

Catalog Number p-7026860
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
719-64-2
C13H8ClNO
229.663
MFCD00014573
MUHJZJKVEQASGY-UHFFFAOYSA-N
5-chloro-3-phenylbenzo c isoxazole, 5-chloro-3-phenylanthranil, 5-chloro-3-phenyl-2,1-benzisoxazole, 3-phenyl-5-chloroanthranil, pubchem8713, acmc-209okx, maybridge1_004397, 2, 5-chloro-3-phenyl, 5-chloro-3-phenyl-benzo c isoxazole, 3-phenyl-5-chloro-2,1-benzisoxazole
347291
5-chloro-3-phenyl-2,1-benzoxazole
C1=CC=C(C=C1)C2=C3C=C(C=CC3=NO2)Cl
This item is not returnable. View return policy
719-64-2
C13H8ClNO
229.663
MFCD00014573
MUHJZJKVEQASGY-UHFFFAOYSA-N
5-chloro-3-phenylbenzo c isoxazole, 5-chloro-3-phenylanthranil, 5-chloro-3-phenyl-2,1-benzisoxazole, 3-phenyl-5-chloroanthranil, pubchem8713, acmc-209okx, maybridge1_004397, 2, 5-chloro-3-phenyl, 5-chloro-3-phenyl-benzo c isoxazole, 3-phenyl-5-chloro-2,1-benzisoxazole
347291
5-chloro-3-phenyl-2,1-benzoxazole
C1=CC=C(C=C1)C2=C3C=C(C=CC3=NO2)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 114°C to 117°C
Quantity 10 g
Formula Weight 229.67
Percent Purity ≥98%
Chemical Name or Material 5-Chloro-3-phenyl-2,1-benzisoxazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'