missing translation for 'onlineSavingsMsg'
Få mere at vide

5-Chloro-3-nitro-pyrazolo[1,5-a]pyrimidine, TRC

Artikelnummer. 30350639
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30350639 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30350639 Leverandør Toronto Research Chemicals Leverandørnr. TRCC371555250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 5-Chloro-3-nitro-pyrazolo[1,5-a]pyrimidine
CAS 1363380-51-1
Molekylær formel C6H3ClN4O2
Mængde 250 mg
Synonym 5-Chloro-3-Nitropyrazolo[1,5-A]Pyrimidine
InChI formel InChI=1S/C6H3ClN4O2/c7-5-1-2-10-6(9-5)4(3-8-10)11(12)13/h1-3H
SMIL [O-][N+](=O)c1cnn2ccc(Cl)nc12
IUPAC navn 5-chloro-3-nitropyrazolo[1,5-a]pyrimidine
Molekylvægt (g/mol) 198.57
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.