missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-3-methylbenzo[b]thiophene, 97%, Thermo Scientific™

Catalog Number p-4813214
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
19404-18-3
C9H7ClS
182.67
UWDQVEPXORTQFO-UHFFFAOYSA-N
5-chloro-3-methylbenzo b thiophene, 5-chloro-3-methylthianaphthene, 5-chloro-3-methyl benzo b thiophene, 5-chloro-3-methylbenzothiophene, 4-methylthio phenol, zlchem 217, pubchem7372, acmc-209exr, maybridge1_005366
317317
5-chloro-3-methyl-1-benzothiophene
CC1=CSC2=C1C=C(C=C2)Cl
This item is not returnable. View return policy
19404-18-3
C9H7ClS
182.67
UWDQVEPXORTQFO-UHFFFAOYSA-N
5-chloro-3-methylbenzo b thiophene, 5-chloro-3-methylthianaphthene, 5-chloro-3-methyl benzo b thiophene, 5-chloro-3-methylbenzothiophene, 4-methylthio phenol, zlchem 217, pubchem7372, acmc-209exr, maybridge1_005366
317317
5-chloro-3-methyl-1-benzothiophene
CC1=CSC2=C1C=C(C=C2)Cl
Melting Point 31.0°C to 32.0°C
Boiling Point 87.0°C (0.5 mmHg)
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 182.67
Percent Purity 97%
Chemical Name or Material 5-Chloro-3-methylbenzo[b]thiophene
compliance-icons
Product Identifier
  • 5-Chloro-3-methylbenzo¢b!thiophene, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'