Få mere at vide
5-Chloro-2-thiazol-4-yl-1H-benzoimidazole, TRC
Artikelnummer. 30361180
Skift visning
Mængde:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30361180 500 mg
30354347 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30361180 Leverandør Toronto Research Chemicals Leverandørnr. TRCC408580500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Chloro-2-thiazol-4-yl-1H-benzoimidazole
CAS 142383-75-3
Molekylær formel C10 H6 Cl N3 S
Mængde 500 mg
Synonym 5-Chloro Tiabendazole
InChI formel InChI=1S/C10H6ClN3S/c11-6-1-2-7-8(3-6)14-10(13-7)9-4-15-5-12-9/h1-5H,(H,13,14)
SMIL Clc1ccc2nc([nH]c2c1)c3cscn3
IUPAC navn 4-(6-chloro-1H-benzimidazol-2-yl)-1,3-thiazole
Molekylvægt (g/mol) 235.693
Formel vægt 234.997