Få mere at vide
5-Chloro-2-nitrodiphenylamine-d5, TRC
Artikelnummer. 30362350
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30362350 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30362350 Leverandør Toronto Research Chemicals Leverandørnr. TRCC37392210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Chloro-2-nitrodiphenylamine-d5
CAS 129973-73-5
Molekylær formel C12H4D5ClN2O2
Mængde 10 mg
Synonym 5-Chloro-2-nitro-N-phenyl-benzenamine-d5, 5-Chloro-2-nitro-diphenylamine-d5, 2-Nitro-5-chlorodiphenylamine-d5, 5-Chloro-2-nitro-N-phenylbenzenamine-d5
InChI formel InChI=1S/C12H9ClN2O2/c13-9-6-7-12(15(16)17)11(8-9)14-10-4-2-1-3-5-10/h1-8,14H/i1D,2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(Nc2cc(Cl)ccc2[N+](=O)[O-])c([2H])c1[2H]
IUPAC navn N-(5-chloro-2-nitrophenyl)-2,3,4,5,6-pentadeuterioaniline
Molekylvægt (g/mol) 253.7
Formel vægt 253.0666