Få mere at vide
5'-Chloro-2'-nitro-N-phenyl-malonanilic Acid-d5 Ethyl Ester, TRC
Artikelnummer. 30359506
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30359506 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30359506 Leverandør Toronto Research Chemicals Leverandørnr. TRCC37432750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5'-Chloro-2'-nitro-N-phenyl-malonanilic Acid-d5 Ethyl Ester
CAS 129973-74-6
Molekylær formel C17H10D5ClN2O5
Mængde 50 mg
Synonym 3-[(5-Chloro-2-nitrophenyl)phenylamino]-3-oxo-propanoic Acid-d5 Ethyl Ester
InChI formel InChI=1S/C17H15ClN2O5/c1-2-25-17(22)11-16(21)19(13-6-4-3-5-7-13)15-10-12(18)8-9-14(15)20(23)24/h3-10H,2,11H2,1H3/i3D,4D,5D,6D,7D
SMIL [2H]c1c([2H])c([2H])c(N(C(=O)CC(=O)OCC)c2cc(Cl)ccc2[N+](=O)[O-])c([2H])c1[2H]
IUPAC navn ethyl 3-(5-chloro-2-nitro-N-(2,3,4,5,6-pentadeuteriophenyl)anilino)-3-oxopropanoate
Molekylvægt (g/mol) 367.8
Formel vægt 367.0983