Learn More
5-chloro-2-nitro-4-(trifluoromethyl)aniline, 97%, Thermo Scientific™
Catalog Number p-2701306
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
35375-74-7
C7H4ClF3N2O2
240.566
MFCD01312277
AAMCWLIPRMIPBN-UHFFFAOYSA-N
5-chloro-2-nitro-4-trifluoromethyl aniline, 4-amino-2-chloro-5-nitrobenzotrifluoride, 5-chloro-2-nitro-4-trifluoromethyl benzenamine, 3-chloro-6-nitro-4-trifluoromethyl aniline, 5-chloro-2-nitro-4-trifluoromethyl-phenylamine, 5-chloro-2-nitro-4-trifluoromethyl phenylamine, 5-chloro-4-trifluoromethyl-2-nitroaniline, benzenamine, 5-chloro-2-nitro-4-trifluoromethyl, 5-chloro-2-nitro-4-trifluoromethylphenyl amine, pubchem2803
2773813
5-chloro-2-nitro-4-(trifluoromethyl)aniline
C1=C(C(=CC(=C1[N+](=O)[O-])N)Cl)C(F)(F)F
35375-74-7
C7H4ClF3N2O2
240.566
MFCD01312277
AAMCWLIPRMIPBN-UHFFFAOYSA-N
5-chloro-2-nitro-4-trifluoromethyl aniline, 4-amino-2-chloro-5-nitrobenzotrifluoride, 5-chloro-2-nitro-4-trifluoromethyl benzenamine, 3-chloro-6-nitro-4-trifluoromethyl aniline, 5-chloro-2-nitro-4-trifluoromethyl-phenylamine, 5-chloro-2-nitro-4-trifluoromethyl phenylamine, 5-chloro-4-trifluoromethyl-2-nitroaniline, benzenamine, 5-chloro-2-nitro-4-trifluoromethyl, 5-chloro-2-nitro-4-trifluoromethylphenyl amine, pubchem2803
2773813
5-chloro-2-nitro-4-(trifluoromethyl)aniline
C1=C(C(=CC(=C1[N+](=O)[O-])N)Cl)C(F)(F)F
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 240.57
Percent Purity 97%
Chemical Name or Material 5-chloro-2-nitro-4-(trifluoromethyl)aniline