Learn More
5-chloro-2-methoxyphenyl isothiocyanate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 199.66 |
Percent Purity | 97% |
Chemical Name or Material | 5-Chloro-2-methoxyphenyl isothiocyanate |