missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methoxybenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-7030957
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
This item is not returnable. View return policy
7035-09-8
MFCD00033854
This item is not returnable. View return policy
7035-09-8
MFCD00033854

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 75°C to 79°C
Odor Characteristic
Molecular Formula C3H10ClNO3S
Quantity 5 g
Sensitivity Air sensitive
Synonym l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
InChI Key QIJRTFXNRTXDIP-JIZZDEOASA-N
SMILES C(C(C(=O)O)N)S.O.Cl
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
Molecular Weight (g/mol) 175.63
PubChem CID 23462
Formula Weight 170.6
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-methoxybenzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'