Learn More
Alfa Aesar™ 5-Chloro-2-methoxybenzaldehyde, 97%
Brand: Alfa Aesar™ H31860.06
Additional Details : CAS Number : 7035-09-8 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
7035-09-8 | |
175.63 | |
QIJRTFXNRTXDIP-JIZZDEOASA-N | |
23462 | |
C(C(C(=O)O)N)S.O.Cl |
C3H10ClNO3S | |
MFCD00033854 | |
l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl | |
(2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride |
Specifications
5-Chloro-2-methoxybenzaldehyde | |
Air sensitive | |
5g | |
C3H10ClNO3S | |
l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl | |
C(C(C(=O)O)N)S.O.Cl | |
175.63 | |
170.6 |
Characteristic | |
75°C to 79°C | |
7035-09-8 | |
MFCD00033854 | |
QIJRTFXNRTXDIP-JIZZDEOASA-N | |
(2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride | |
23462 | |
97% |