missing translation for 'onlineSavingsMsg'
Få mere at vide

5-Chloro-2-methoxy-benzoic Acid Methyl Ester-13C2,d6, TRC

Artikelnummer. 30361717
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30361717 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361717 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36790750MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 5-Chloro-2-methoxy-benzoic Acid Methyl Ester-13C2,d6
CAS 1794737-29-3
Molekylær formel C713C2H3D6ClO3
Mængde 50 mg
Synonym 5-Chloro-o-anisic Acid Methyl Ester-13C2,d6, 4-Chloro-2-(methoxycarbonyl)anisole-13C2,d6, 5-Chloro-2-methoxybenzoic Acid Methyl Ester-13C2,d6, Methyl 2-Methoxy-5-chlorobenzoate-13C2,d6, Methyl 5-Chloro-2-methoxybenzoate-13C2,d6
InChI formel InChI=1S/C9H9ClO3/c1-12-8-4-3-6(10)5-7(8)9(11)13-2/h3-5H,1-2H3/i1+1D3,2+1D3
SMIL [2H][13C]([2H])([2H])OC(=O)c1cc(Cl)ccc1O[13C]([2H])([2H])[2H]
IUPAC navn trideuterio(113C)methyl 5-chloro-2-(trideuterio(113C)methoxy)benzoate
Molekylvægt (g/mol) 208.64
Formel vægt 208.0684
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.