Få mere at vide
5-Chloro-2-methoxy-benzoic Acid-13C,d3, TRC
Artikelnummer. 30357395
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30357395 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30357395 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3679122.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Chloro-2-methoxy-benzoic Acid-13C,d3
CAS 1346600-36-9
Molekylær formel C713CH4D3ClO3
Mængde 2.5 mg
Synonym 5-Chloro-o-anisic-13C,d3 Acid, 2-Methoxy-13C,d3-5-chlorobenzoic Acid, 5-Chloro-2-methoxy-13C,d3-benzoic Acid, 5-Chloro-o-anisic-13C,d3 Acid
InChI formel InChI=1S/C8H7ClO3/c1-12-7-3-2-5(9)4-6(7)8(10)11/h2-4H,1H3,(H,10,11)/i1+1D3
SMIL [2H][13C]([2H])([2H])Oc1ccc(Cl)cc1C(=O)O
IUPAC navn 5-chloro-2-(trideuterio(113C)methoxy)benzoic acid
Molekylvægt (g/mol) 190.6
Formel vægt 190.0306