Learn More
5-Chloro-2-iodopyrimidine, 98%, Thermo Scientific™
Catalog Number p-7055573
Click to view available options
Quantity:
250 mg
1 g
5 g
874676-81-0
C4H2ClIN2
240.428
MFCD08062390
BWDLBDLGVMLSCS-UHFFFAOYSA-N
pyrimidine, 5-chloro-2-iodo, pubchem14957, acmc-209qnq, 5-chloro-2-iodo pyrimidine, ksc493q5d
29938005
5-chloro-2-iodopyrimidine
C1=C(C=NC(=N1)I)Cl
874676-81-0
C4H2ClIN2
240.428
MFCD08062390
BWDLBDLGVMLSCS-UHFFFAOYSA-N
pyrimidine, 5-chloro-2-iodo, pubchem14957, acmc-209qnq, 5-chloro-2-iodo pyrimidine, ksc493q5d
29938005
5-chloro-2-iodopyrimidine
C1=C(C=NC(=N1)I)Cl
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Useful Intermediate for organic synthesis and pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Store in dark. Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Beilstein 14516398
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 240.43
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Chloro-2-iodopyrimidine