missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-2-iodopyridine, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is applied in an efficient preparation of indolizines through a tandem palladium-catalyzed cross-coupling reaction and cycloisomerization and also in the synthesis, and structure-activity relationships of 2-aminoquinazoline derivatives as a novel class of metabotropic glutamate receptor 5 negative allosteric modulators.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Store away from heat, light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from exposure to light. Keep it refrigerated.
It is applied in an efficient preparation of indolizines through a tandem palladium-catalyzed cross-coupling reaction and cycloisomerization and also in the synthesis, and structure-activity relationships of 2-aminoquinazoline derivatives as a novel class of metabotropic glutamate receptor 5 negative allosteric modulators.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Store away from heat, light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from exposure to light. Keep it refrigerated.
Specifications
Quantity | 1 g |
Beilstein | 9321405 |
Sensitivity | Light sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 239.44 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 5-Chloro-2-iodopyridine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'