Få mere at vide
5-Chloro-2-furancarbonyl Chloride (~ 90%), TRC
Artikelnummer. 30348925
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30348925 100 mg
30365855 500 mg
30367767 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30348925 Leverandør Toronto Research Chemicals Leverandørnr. TRCC376265100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 24078-79-3
Molekylær formel C5H2Cl2O2
Molekylvægt (g/mol) 164.97
Synonym 5-Chloro-2-furoyl Chloride
IUPAC navn 5-chlorofuran-2-carbonyl chloride
SMIL O=C(Cl)C1=CC=C(Cl)O1
Tekniske data
Kemisk navn eller materiale 5-Chloro-2-furancarbonyl Chloride
Mængde 100 mg
InChI formel InChI=1S/C5H2Cl2O2/c6-4-2-1-3(9-4)5(7)8/h1-2H
Formel vægt 163.9432