missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloro-2-(3-methoxyphenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64017.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15471029

  • 2403.83 DKK / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

5-Chloro-2-(3-methoxyphenyl)benzimidazole
258.7
MFCD20043307

Specifications

5-Chloro-2-(3-methoxyphenyl)benzimidazole
Solid
258.7
1g
95%
MFCD20043307
Videos
SDS