Få mere at vide
5-Carboxyvanillin, TRC
Artikelnummer. 30349915
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30349915 500 mg
30359699 1 g
30356901 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30349915 Leverandør Toronto Research Chemicals Leverandørnr. TRCC183318500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3507-08-2
Molekylær formel C9H8O5
Molekylvægt (g/mol) 196.16
Synonym 5-Formyl-3-methoxysalicylic Acid, 5-Formyl-o-vanillic Acid, 5-Formyl-2-hydroxy-3-methoxybenzoic Acid, 3-Carboxy-4-hydroxy-5-methoxybenzaldehyde, 5-Carboxy-4-hydroxy-3-methoxybenzaldehyde
IUPAC navn 5-formyl-2-hydroxy-3-methoxybenzoic acid
SMIL COc1cc(C=O)cc(C(=O)O)c1O
Tekniske data
Kemisk navn eller materiale 5-Carboxyvanillin
Mængde 500 mg
InChI formel InChI=1S/C9H8O5/c1-14-7-3-5(4-10)2-6(8(7)11)9(12)13/h2-4,11H,1H3,(H,12,13)
Formel vægt 196.0372