missing translation for 'onlineSavingsMsg'
Learn More

5-Carbethoxyuracil, 98%, Thermo Scientific™

Catalog Number p-3735919
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
28485-17-8
C7H8N2O4
184.15
MFCD00057337
MKNYHTGOVKPZMU-UHFFFAOYSA-N
5-carbethoxyuracil, ethyl4-hydroxy-2-oxo-1,2-dihydropyrimidine-5-carboxylate, ethyl 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate, isoorotic acid ethyl ester, ethyl 2,4-dihydroxypyrimidine-5-carboxylate, ethyl uracil-5-carboxylate, ethyl 1,2,3,4-tetrahydro-2,4-dioxopyrimidine-5-carboxylate, 5-pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-2,4-dioxo-, ethyl ester, ethyl 2,4-dioxo-1,3-dihydropyrimidine-5-carboxylate, pubchem13254
101411
ethyl 2,4-dioxo-1H-pyrimidine-5-carboxylate
CCOC(=O)C1=CNC(=O)NC1=O
This item is not returnable. View return policy
28485-17-8
C7H8N2O4
184.15
MFCD00057337
MKNYHTGOVKPZMU-UHFFFAOYSA-N
5-carbethoxyuracil, ethyl4-hydroxy-2-oxo-1,2-dihydropyrimidine-5-carboxylate, ethyl 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate, isoorotic acid ethyl ester, ethyl 2,4-dihydroxypyrimidine-5-carboxylate, ethyl uracil-5-carboxylate, ethyl 1,2,3,4-tetrahydro-2,4-dioxopyrimidine-5-carboxylate, 5-pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-2,4-dioxo-, ethyl ester, ethyl 2,4-dioxo-1,3-dihydropyrimidine-5-carboxylate, pubchem13254
101411
ethyl 2,4-dioxo-1H-pyrimidine-5-carboxylate
CCOC(=O)C1=CNC(=O)NC1=O
CAS Min % 97.5
CAS Max % 100.0
Melting Point 232.0°C to 235.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 184.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Carbethoxyuracil
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'