missing translation for 'onlineSavingsMsg'
Learn More

5-Bromopicolinic acid, 97%, ACROS Organics™

Brand:  Acros Organics 393990010

Additional Details : CAS Number : 30766-11-1 Weight : 0.05100kg


Packaging Glass bottle
Quantity 1g

 View more versions of this product

Product Code. 11986331

  • 163.00 DKK / 1g

Estimated Shipment
04-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

30766-11-1
202.01
5-bromopicolinic acid, 5-bromo-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-bromo, 5-bromo-2-carboxypyridine, 5-bromo-pyridine-2-carboxylic acid, 5-bromo-2-picolinic acid, 5-bromo-2-carboxy pyridine, 5-bromo-2-pyridine carboxylic acid, 5-bromopyridine-2-carboxylicacid
5-bromopyridine-2-carboxylic acid
C6H4BrNO2
MNNQIBXLAHVDDL-UHFFFAOYSA-N
608544
C1=CC(=NC=C1Br)C(=O)O

Specifications

5-Bromopicolinic acid
Glass bottle
177.0°C to 178.0°C
30766-11-1
100.0
C6H4BrNO2
MNNQIBXLAHVDDL-UHFFFAOYSA-N
5-bromopyridine-2-carboxylic acid
608544
Powder
Authentic
White
1g
96.0
96% min. (HPLC)
5-bromopicolinic acid, 5-bromo-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-bromo, 5-bromo-2-carboxypyridine, 5-bromo-pyridine-2-carboxylic acid, 5-bromo-2-picolinic acid, 5-bromo-2-carboxy pyridine, 5-bromo-2-pyridine carboxylic acid, 5-bromopyridine-2-carboxylicacid
C1=CC(=NC=C1Br)C(=O)O
202.01
202.01
97%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Bromopicolinic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents