Få mere at vide
5-Bromopentanoic-3,3,4,4-d4 Acid, TRC
Artikelnummer. 30333838
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30333838 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30333838 Leverandør Toronto Research Chemicals Leverandørnr. TRCB686147100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Bromopentanoic-3,3,4,4-d4 Acid
CAS 42932-62-7
Molekylær formel C5 D4 H5 Br O2
Mængde 100 mg
Synonym 5-Bromovaleric Acid-d4, 1-Bromo-4-carboxybutane-d4, 5-Bromo-1-pentanoic Acid-d4, 5-Bromopentanoic Acid-d4, 5-Bromovaleric Acid-d4, NSC 53507-d4, delta-Bromovaleric Acid-d4
InChI formel InChI=1S/C5H9BrO2/c6-4-2-1-3-5(7)8/h1-4H2,(H,7,8)/i1D2,2D2
SMIL [2H]C([2H])(CBr)C([2H])([2H])CC(=O)O
IUPAC navn 5-bromo-3,3,4,4-tetradeuteriopentanoic acid
Molekylvægt (g/mol) 185.05
Formel vægt 184.0037