Få mere at vide
5-Bromonicotinic acid, 98%, Thermo Scientific™
Artikelnummer. 11420124
Skift visning
Mængde:
5 g
25 g
100 g
Pakningsstørrelse:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11420124 5 g 5g
11430124 25 g 25g
11440124 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 11420124 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. A13589.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 20826-04-4
Molekylær formel C6H4BrNO2
Molekylvægt (g/mol) 202.01
MDL nummer MFCD00009783
InChI nøgle FQIUCPGDKPXSLL-UHFFFAOYSA-N
Synonym 5-bromonicotinic acid, 5-bromo nicotinic acid, 5-bromo-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-bromo, nicotinic acid, 5-bromo, 5-bromo-nicotinic acid, 3-bromo-5-pyridine carboxylic acid, 3-bromo pyridine-5-carboxylic acid, pubchem1211, acmc-209fev
PubChem CID 88707
IUPAC navn 5-bromopyridine-3-carboxylic acid
SMIL OC(=O)C1=CN=CC(Br)=C1
Tekniske data
Kemisk navn eller materiale 5-Bromonicotinic acid
Smeltepunkt 182°C to 186°C
Mængde 5 g
Beilstein 115854
Formel vægt 202.01
Procent renhed 98%